Anti-NR2C2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
USBI039118-APCEA 852 EUR
USBI039118-APC
Anti-NR2C2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: NR2C2 (nuclear receptor subfamily 2, group C, member 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human