Anti-NSUN2 Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI039167EA 587 EUR
USBI039167
Anti-NSUN2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NSUN2 (NOP2/Sun RNA methyltransferase family, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR