Anti-OGN Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
USBI039270-APCEA 852 EUR
USBI039270-APC
Anti-OGN Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: OGN (osteoglycin)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human