Anti-SBK3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: United States Biological
USBI041656-APCEA 852 EUR
USBI041656-APC
Anti-SBK3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SBK3 (SH3 domain binding kinase family, member 3)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse