Anti-Sgk223 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: United States Biological
USBI041660-FITCEA 852 EUR
USBI041660-FITC
Anti-Sgk223 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: Sgk223
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR