Anti-ELP3 Rabbit polyclonal antibody

Lieferant: United States Biological
USBIE2249-82AEA 587 EUR
USBIE2249-82A
Anti-ELP3 Rabbit polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: ELP3 (elongator acetyltransferase complex subunit 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR