Anti-ELOVL6 Rabbit polyclonal antibody

Lieferant: United States Biological
USBIE2251-12EA 587 EUR
USBIE2251-12
Anti-ELOVL6 Rabbit polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: ELOVL6 (ELOVL fatty acid elongase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR