Anti-ELOVL7 Rabbit polyclonal antibody

Lieferant: United States Biological
USBIE2251-13EA 633 EUR
USBIE2251-13
Anti-ELOVL7 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: ELOVL7 (ELOVL fatty acid elongase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR