Anti-EM-66 Rabbit polyclonal antibody

Lieferant: United States Biological
USBIE2256-26AEA 979 EUR
USBIE2256-26A
Anti-EM-66 Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: EM-66
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR