Anti-EML2 Goat polyclonal antibody

Lieferant: United States Biological
USBIE2258-10DEA 633 EUR
USBIE2258-10D
Anti-EML2 Goat polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: EML2 (echinoderm microtubule associated protein like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR