Anti-EMP3 Mouse monoclonal antibody [clone: 8B16]

Lieferant: United States Biological
USBIE2261EA 742 EUR
USBIE2261
Anti-EMP3 Mouse monoclonal antibody [clone: 8B16]
Antikörper
Type: Primary
Antigen: EMP3 (epithelial membrane protein 3)
Clonality: monoclonal
Clone: 8B16
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR