Anti-EN2, CT-2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: United States Biological
USBIE2270-10C-HRPEA 852 EUR
USBIE2270-10C-HRP
Anti-EN2, CT-2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: EN2, CT-2
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human