Anti-Nucleoside Diphosphate Kinase 6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: United States Biological
USBIN7100-68N-APCEA 852 EUR
USBIN7100-68N-APC
Anti-Nucleoside Diphosphate Kinase 6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: Nucleoside Diphosphate Kinase 6
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR