Anti-Nucleoside Diphosphate Kinase 7 Rabbit Polyclonal Antibody (Biotin)
Lieferant: United States Biological
USBIN7100-68T-BIOTEA 852 EUR
USBIN7100-68T-BIOT
Anti-Nucleoside Diphosphate Kinase 7 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Nucleoside Diphosphate Kinase 7
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human