Anti-Nucleoside Monophosphate Kinase Rabbit Polyclonal Antibody
Lieferant: United States Biological
USBIN7100-69AEA 809 EUR
USBIN7100-69A
Anti-Nucleoside Monophosphate Kinase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Nucleoside Monophosphate Kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen