Anti-Nucleoside Monophosphate Kinase Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIN7100-69BEA 608 EUR
USBIN7100-69B
Anti-Nucleoside Monophosphate Kinase Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Nucleoside Monophosphate Kinase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR