Anti-NT5E Mouse Monoclonal Antibody [clone: 9G246]

Lieferant: United States Biological
USBIN7100-71CEA 723 EUR
USBIN7100-71C
Anti-NT5E Mouse Monoclonal Antibody [clone: 9G246]
Antikörper
Type: Primary
Antigen: NT5E (5'-nucleotidase, ecto (CD73))
Clonality: monoclonal
Clone: 9G246
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR