Anti-NUAK2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIN7400-10EA 587 EUR
USBIN7400-10
Anti-NUAK2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: NUAK2 (NUAK family, SNF1-like kinase, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR