Anti-NUAK2 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: United States Biological
USBIN7400-10-FITCEA 852 EUR
USBIN7400-10-FITC
Anti-NUAK2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: NUAK2 (NUAK family, SNF1-like kinase, 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR