Anti-OSM Rabbit Polyclonal Antibody (Biotin)
Lieferant: United States Biological
USBIO6500-10CEA 668 EUR
USBIO6500-10C
Anti-OSM Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 0.15-0. 3?g/ml; Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: OSM (oncostatin M)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human