Anti-OSM Mouse Monoclonal Antibody [clone: 12K146]

Lieferant: United States Biological
USBIO6500-10DEA 979 EUR
USBIO6500-10D
Anti-OSM Mouse Monoclonal Antibody [clone: 12K146]
Antikörper
Type: Primary
Antigen: OSM (oncostatin M)
Clonality: monoclonal
Clone: 12K146
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR