Anti-OSMR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 10K58]

Lieferant: United States Biological
USBIO6500-50AEA 776 EUR
USBIO6500-50A
Anti-OSMR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 10K58]
Antikörper
Do not freeze!

Type: Primary
Antigen: OSMR (oncostatin M receptor)
Clonality: monoclonal
Clone: 10K58
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR