Anti-OPA1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIO7004-85EA 574 EUR
USBIO7004-85
Anti-OPA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OPA1 (optic atrophy 1 (autosomal dominant))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR