Anti-Opioid Receptor, delta Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIO7030-15BEA 634 EUR
USBIO7030-15B
Anti-Opioid Receptor, delta Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Opioid Receptor, delta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR