Anti-OPRK1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIO7030-25AEA 641 EUR
USBIO7030-25A
Anti-OPRK1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: OPRK1 (opioid receptor, kappa 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR