Anti-OPRK1 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIO7030-25BEA 551 EUR
USBIO7030-25B
Anti-OPRK1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: OPRK1 (opioid receptor, kappa 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR