Anti-Opioid Receptor, kappa Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIO7030-31AEA 410 EUR
USBIO7030-31A
Anti-Opioid Receptor, kappa Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1500

Type: Primary
Antigen: Opioid Receptor, kappa
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR