Anti-Opioid Receptor, mu Rabbit polyclonal antibody

USBIO7030-35AEA 454 EUR
USBIO7030-35A
Anti-Opioid Receptor, mu Rabbit polyclonal antibody
Recommended dilution: Immunochemistry: 1:2000; Western Blot: 1:500

Type: Primary
Antigen: Opioid Receptor, mu
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR