Anti-Opioid Receptor 1, mu Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIO7030-39DEA 672 EUR
USBIO7030-39D
Anti-Opioid Receptor 1, mu Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:100; Immunoprecipitation: 1:100

Type: Primary
Antigen: Opioid Receptor 1, mu
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR