Anti-OPRM1 Mouse Monoclonal Antibody [clone: 12K148]

Lieferant: United States Biological
USBIO7030-39FEA 748 EUR
USBIO7030-39F
Anti-OPRM1 Mouse Monoclonal Antibody [clone: 12K148]
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: OPRM1 (opioid receptor, mu 1)
Clonality: monoclonal
Clone: 12K148
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR