Anti-Opioid Receptor, mu Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIO7030-50AEA 410 EUR
USBIO7030-50A
Anti-Opioid Receptor, mu Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:5000; Immunochemistry: 1:2000

Type: Primary
Antigen: Opioid Receptor, mu
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR