Anti-OPTC Rat Monoclonal Antibody [clone: 7H137]

Lieferant: United States Biological
USBIO7034-50EA 729 EUR
USBIO7034-50
Anti-OPTC Rat Monoclonal Antibody [clone: 7H137]
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: OPTC (opticin)
Clonality: monoclonal
Clone: 7H137
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR