Anti-OPTN Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIO7037EA 552 EUR
USBIO7037
Anti-OPTN Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: OPTN (optineurin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR