Anti-ORAI1 Rabbit polyclonal antibody

USBIO7044-03BEA 352 EUR
USBIO7044-03B
Anti-ORAI1 Rabbit polyclonal antibody
Recommended dilution: Western Blot: 1:500-1:1000; ELISA: 1:5000-1:20000

Type: Primary
Antigen: ORAI1 (ORAI calcium release-activated calcium modulator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR