Anti-Presenilin Enhancer 2 Rabbit Polyclonal Antibody
Lieferant: United States Biological
USBIP6300-80DEA 532 EUR
USBIP6300-80D
Anti-Presenilin Enhancer 2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: Presenilin Enhancer 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat