Anti-PSENEN Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIP6300-81DEA 633 EUR
USBIP6300-81D
Anti-PSENEN Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: PSENEN (presenilin enhancer gamma secretase subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR