Anti-POFUT1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIP6401-01EA 526 EUR
USBIP6401-01
Anti-POFUT1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: POFUT1 (protein O-fucosyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR