Anti-PRG4 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIP8930-03AEA 587 EUR
USBIP8930-03A
Anti-PRG4 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:128000

Type: Primary
Antigen: PRG4 (proteoglycan 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR