Anti-Prickle-1b Rabbit polyclonal antibody
USBIP8947-01WEA 319 EUR
USBIP8947-01W
Anti-Prickle-1b Rabbit polyclonal antibody
Recommended dilution: ELISA: 1:5000-1:20000
Type: Primary
Antigen: Prickle-1b
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish