Anti-PRNP Mouse Monoclonal Antibody [clone: 9H266]
Lieferant: United States Biological
USBIP8950-01AEA 590 EUR
USBIP8950-01A
Anti-PRNP Mouse Monoclonal Antibody [clone: 9H266]
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 1:50-1:300
Type: Primary
Antigen: PRNP (prion protein)
Clonality: monoclonal
Clone: 9H266
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Rat