Anti-PRNP Mouse Monoclonal Antibody [clone: EM-20]
Lieferant: United States Biological
USBIP8950-26DEA 675 EUR
USBIP8950-26D
Anti-PRNP Mouse Monoclonal Antibody [clone: EM-20]
Antikörper
Recommended dilution: Immunochemistry: 10 ?g/ml; Western Blot: 0.5 ?g/ml
Type: Primary
Antigen: PRNP (prion protein)
Clonality: monoclonal
Clone: EM-20
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human