Anti-PRKDC Rabbit Polyclonal Antibody (Biotin)
Lieferant: United States Biological
USBIP9003-01C-BIOTEA 852 EUR
USBIP9003-01C-BIOT
Anti-PRKDC Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: PRKDC (protein kinase, DNA-activated, catalytic polypeptide)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human