Anti-PRKDC Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: United States Biological
USBIP9003-01F-PEEA 852 EUR
USBIP9003-01F-PE
Anti-PRKDC Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: PRKDC (protein kinase, DNA-activated, catalytic polypeptide)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human