Anti-PRKDC Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIP9003-01LEA 568 EUR
USBIP9003-01L
Anti-PRKDC Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 2?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: PRKDC (protein kinase, DNA-activated, catalytic polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR