Anti-PRMT1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIP9004-05A-BIOTEA 852 EUR
USBIP9004-05A-BIOT
Anti-PRMT1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: PRMT1 (protein arginine methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR