Anti-PRMT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: United States Biological
USBIP9004-05A-HRPEA 852 EUR
USBIP9004-05A-HRP
Anti-PRMT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: PRMT1 (protein arginine methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR