Anti-PRMT1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIP9004-05DEA 633 EUR
USBIP9004-05D
Anti-PRMT1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:50-1:100; Western Blot: 1:50-1:100; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: PRMT1 (protein arginine methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR