Anti-PRL3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: United States Biological
USBIP9109-48C3-APCEA 852 EUR
USBIP9109-48C3-APC
Anti-PRL3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: PRL3
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse