Anti-PRL3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Lieferant: United States Biological
USBIP9109-48C3FITCEA 852 EUR
USBIP9109-48C3FITC
Anti-PRL3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: PRL3
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse