Anti-PRL3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: United States Biological
USBIP9109-48C3-HRPEA 852 EUR
USBIP9109-48C3-HRP
Anti-PRL3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: PRL3
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse